Learn More
Cayman Chemical MethyclothIazIde 50mg
SDP
Catalog No. 502994928
Encompass_Preferred
A thiazide diuretic; inhibits norepinephrine- and vasopressin-induced contractions of aortic rings isolated from spontaneously hypertensive rats at a concentration of 100 μM; reduces salt-induced increases in blood pressure in salt-sensitive but not salt-resistant rats without affecting body weight or heart rate
Catalog No. 50-299-4928 Supplier Cayman Chemical Supplier No. 2379950 MG
A thiazide diuretic; inhibits norepinephrine- and vasopressin-induced contractions of aortic rings isolated from spontaneously hypertensive rats at a concentration of 100 μM; reduces salt-induced increases in blood pressure in salt-sensitive but not salt-resistant rats without affecting body weight or heart rate