Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1,2,4-triazole-3-carboxylate, 98%

Catalog No. AC415000050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC415000050 5 g
AC415000250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC415000050 Supplier Thermo Scientific Chemicals Supplier No. 415000050
Add to Cart

CAS: 4928-88-5 | C4H5N3O2 | 127.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4928-88-5
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.1
InChI Key QMPFMODFBNEYJH-UHFFFAOYSA-N
PubChem CID 2735089
IUPAC Name methyl 1H-1,2,4-triazole-5-carboxylate
SMILES COC(=O)C1=NC=NN1

Specifications

Melting Point 190.0°C to 191.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Synonym methyl 1h-1,2,4-triazole-3-carboxylate, methyl 1,2,4-triazole-3-carboxylate, 1h-1,2,4-triazole-3-carboxylic acid methyl ester, methyl 4h-1,2,4-triazole-3-carboxylate, 1,2,4-triazole-3-carboxylic acid methylester, 1h-1,2,4-triazole-3-carboxylic acid, methyl ester, 1h-1,2,4-triazole-5-carboxylic acid methyl ester, methyl1,2,4-triazole-3-carboxylate, acmc-20a2ir, 5-carbomethoxy-1,2,4-triazole
Formula Weight 127.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 1,2,4-triazole-3-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.