Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-Adamantanecarboxylate 98.0+%, TCI America™
SDP

Catalog No. M211925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M211925G 25 g
M21195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M211925G Supplier TCI America Supplier No. M211925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 711-01-3
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.274
MDL Number MFCD01838519
InChI Key CLYOOVNORYNXMD-UHFFFAOYSA-N
Synonym 1-Adamantanecarboxylic Acid Methyl Ester
PubChem CID 136553
IUPAC Name methyl adamantane-1-carboxylate
SMILES COC(=O)C12CC3CC(C1)CC(C3)C2

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 79°C
Quantity 25 g
Formula Weight 194.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-Adamantanecarboxylate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.