Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-aminocyclobutanecarboxylate hydrochloride, 95%, Thermo Scientific™

Catalog No. AAH64701MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64701MD 250 mg
AAH6470103 1 g
AAH6470106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64701MD Supplier Thermo Scientific Chemicals Supplier No. H64701MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as a primary and secondary intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92398-47-5
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD01318265
InChI Key SGXOPTPGZBLQAY-UHFFFAOYSA-N
Synonym methyl 1-aminocyclobutanecarboxylate, 1-amino-cyclobutanecarboxylic acid methyl ester, 1-aminocyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, 1-amino-, methyl ester, 1-amino-cyclobutanecarboxylicacidmethylester, methyl 1-amino-cyclobutane-1-carboxylate, 1-aminocyclobutane-1-carboxylic acid methyl ester, cyclobutanecarboxylicacid, 1-amino-, methyl ester
PubChem CID 1502033
IUPAC Name methyl 1-aminocyclobutane-1-carboxylate
SMILES COC(=O)C1(N)CCC1

Specifications

Quantity 250 mg
Beilstein 5615121
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 165.62
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 1-aminocyclobutanecarboxylate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.