Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-Aminocyclopentanecarboxylate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M23515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23515G 5 g
M23511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23515G Supplier TCI America Supplier No. M23515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60421-23-0
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.64
MDL Number MFCD03840364
InChI Key OPUJUITUYWGUEP-UHFFFAOYSA-N
Synonym 1-Aminocyclopentanecarboxylic Acid Methyl Ester Hydrochloride, Cycloleucine Methyl Ester Hydrochloride
PubChem CID 11513856
IUPAC Name hydrogen methyl 1-aminocyclopentane-1-carboxylate chloride
SMILES [H+].[Cl-].COC(=O)C1(N)CCCC1

Specifications

Melting Point 203°C
Color White
Quantity 5 g
Formula Weight 179.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-Aminocyclopentanecarboxylate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.