Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M25315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25315G 5 g
M253125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25315G Supplier TCI America Supplier No. M25315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3939-01-3
Molecular Formula C14H18ClNO3
Molecular Weight (g/mol) 283.75
MDL Number MFCD00012799
InChI Key BRADBAOVPACOQQ-UHFFFAOYNA-N
Synonym methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5, ksc790s5l
PubChem CID 107479
IUPAC Name methyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
SMILES COC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl

Specifications

Melting Point 182°C
Quantity 5 g
Formula Weight 283.75
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.