Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 95%

Catalog No. AAH2693014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2693014 25 g
AAH2693022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2693014 Supplier Thermo Scientific Chemicals Supplier No. H2693014
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 3939-01-3 | C7H3F2N | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3939-01-3
MDL Number MFCD00012799

Specifications

Melting Point ∽185°C (decomposition)
Molecular Formula C14H18ClNO3
Quantity 25 g
Sensitivity Hygroscopic
Synonym methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5, ksc790s5l
InChI Key BRADBAOVPACOQQ-UHFFFAOYNA-N
SMILES COC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl
IUPAC Name methyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
Molecular Weight (g/mol) 283.75
PubChem CID 107479
Formula Weight 283.75
Percent Purity 95%
Chemical Name or Material Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Show More Show Less
EINECSNumber 223-522-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.