Learn More
Medchemexpress LLC Methyl 1-Boc-azetidi | 10g
SDP
Catalog No. 5000203201
Encompass_Preferred
Methyl 1-Boc-azetidine-3-carboxylate is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) Methyl 1-Boc-azetidine-3-carboxylate is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[1
Catalog No. 50-002-03201 Supplier Medchemexpress LLC Supplier No. HY4015110G
Methyl 1-Boc-azetidine-3-carboxylate is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) Methyl 1-Boc-azetidine-3-carboxylate is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs[1