Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 1-Cbz-azetidine-3-carboxylate | 757239-60-4 | MFCD11110708 | 99.6% | 249.26 g/mol | C13H15NO4 | 10 G
SDP

Catalog No. 5000203135
Encompass_Preferred

Methyl 1-Cbz-azetidine-3-carboxylate is a protected azetidine-derived building block used as a non-cleavable antibody-drug conjugate (ADC) linker and as an alkyl-chain PROTAC linker in medicinal chemistry and bioconjugation. The compound is supplied for research use only.

  • High purity 99.6% (GC).
  • Molecular weight 249.26 g/mol; chemical formula C13H15NO4.
  • Suitable for ADC and PROTAC linker synthesis.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Provided for research use only; not for human use.

Catalog No. 50-002-03135 Supplier Medchemexpress LLC Supplier No. HYW01922610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 1-Cbz-azetidine-3-carboxylate is a protected azetidine-derived building block used as a non-cleavable antibody-drug conjugate (ADC) linker and as an alkyl-chain PROTAC linker in medicinal chemistry and bioconjugation. The compound is supplied for research use only.

  • High purity 99.6% (GC).
  • Molecular weight 249.26 g/mol; chemical formula C13H15NO4.
  • Suitable for ADC and PROTAC linker synthesis.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Provided for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.