Learn More
Accela Chembio Inc Methyl 1-methylimidazole-5-carboxylate | 5g | 17289-20-2 | MFCD01567300 | 97+% | Shelf Life: 720 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50251340
Encompass_Preferred
Methyl 1-methylimidazole-5-carboxylate | 5g | 17289-20-2 | MFCD01567300 | 97+% | Shelf Life: 720 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-251-340 Supplier Accela Chembio Inc Supplier No. SY0076525G
Add to Cart
Add to Cart
Methyl 1-methylimidazole-5-carboxylate | 5g | 17289-20-2 | MFCD01567300 | 97+% | Shelf Life: 720 Days | Light Sensitive/nitrogen Or Argon/+4