Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-Naphthoate 98.0+%, TCI America™
SDP

Catalog No. N061525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N061525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N061525G Supplier TCI America Supplier No. N061525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2459-24-7
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00094243
InChI Key HMRROBKAACRWBP-UHFFFAOYSA-N
Synonym Methyl 1-Naphthalenecarboxylate, 1-Naphthoic Acid Methyl Ester, 1-Naphthalenecarboxylic Acid Methyl Ester
PubChem CID 120056
IUPAC Name methyl naphthalene-1-carboxylate
SMILES COC(=O)C1=CC=CC2=CC=CC=C21

Specifications

Color Yellow
Boiling Point 169°C
Quantity 25 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 1-Naphthoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.