Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate 98.0+%, TCI America™
SDP

Catalog No. M291825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M291825G 25 g
M29185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M291825G Supplier TCI America Supplier No. M291825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 148763-41-1
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD06795926
InChI Key LYYJQMCPEHXOEW-UHFFFAOYSA-N
Synonym Methyl 1-Boc-3-piperidinecarboxylate, 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic Acid Methyl Ester, 1-Boc-3-piperidinecarboxylic Acid Methyl Ester, 1-tert-Butyl 3-Methyl Piperidine-1,3-dicarboxylate, Piperidine-1,3-dicarboxylic Acid 1-tert-Butyl 3-Met
PubChem CID 23590652
IUPAC Name 1-O-tert-butyl 3-O-methyl piperidine-1,3-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(=O)OC

Specifications

Melting Point 49°C
Color White-Yellow
Quantity 25 g
Formula Weight 243.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.