Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 11-Bromoundecanoate 97.0+%, TCI America™
SDP

Catalog No. M24125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24125G Supplier TCI America Supplier No. M24125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6287-90-7
Molecular Formula C12H23BrO2
Molecular Weight (g/mol) 279.218
MDL Number MFCD00045047
InChI Key HFNPVFKUZYCDIB-UHFFFAOYSA-N
Synonym 11-Bromoundecanoic Acid Methyl Ester
PubChem CID 223913
IUPAC Name methyl 11-bromoundecanoate
SMILES COC(=O)CCCCCCCCCCBr

Specifications

Melting Point 13°C
Color Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 279.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 11-Bromoundecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.