Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 12-aminododecanoate hydrochloride | 4271-86-7 | 97.0% | 265.82 g/mol | C13H28ClNO2 | 250 MG
SDP

Catalog No. 5000208477
Encompass_Preferred

Methyl 12-aminododecanoate hydrochloride is the hydrochloride salt of methyl 12-aminododecanoate supplied as a solid biochemical reagent for laboratory research. It is used as a reagent, an intermediate in organic synthesis, and for biochemical assay development and reagent preparation.

  • Biochemical reagent for research use.
  • Hydrochloride salt, solid form.
  • Molecular weight 265.82 g/mol.
  • CAS number 4271-86-7.
  • Typical pack sizes include 250 mg, 1 g, and 5 g.
  • Reported purity approximately 97.0%.

Catalog No. 50-002-08477 Supplier Medchemexpress LLC Supplier No. 250MGHYW068215A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 12-aminododecanoate hydrochloride is the hydrochloride salt of methyl 12-aminododecanoate supplied as a solid biochemical reagent for laboratory research. It is used as a reagent, an intermediate in organic synthesis, and for biochemical assay development and reagent preparation.

  • Biochemical reagent for research use.
  • Hydrochloride salt, solid form.
  • Molecular weight 265.82 g/mol.
  • CAS number 4271-86-7.
  • Typical pack sizes include 250 mg, 1 g, and 5 g.
  • Reported purity approximately 97.0%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.