Learn More
Medchemexpress LLC Methyl 1H-1,2,4-triazole-5-carboxylate | 4928-88-5 | 127.10 | 100 G
SDP
Catalog No. 5000446418
Encompass_Preferred
Methyl 1H-1,2,4-triazole-5-carboxylate is a biochemical reagent suitable for life science related research. It is designed for research use only and is not sold to patients. This product presents as a white to off-white solid.
  • Biochemical reagent for life science research
  • Solid appearance
  • White to off-white color
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-46418 Supplier Medchemexpress LLC Supplier No. HYW007685100G
Methyl 1H-1,2,4-triazole-5-carboxylate is a biochemical reagent suitable for life science related research. It is designed for research use only and is not sold to patients. This product presents as a white to off-white solid.
  • Biochemical reagent for life science research
  • Solid appearance
  • White to off-white color
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month