Learn More
Medchemexpress LLC Methyl 1H-pyrrole-2-carboxylate | 1193-62-0 | 99.98% | 125.13 | 25 G
SDP
Catalog No. 5000452441
Encompass_Preferred
Methyl 1H-pyrrole-2-carboxylate is a drug intermediate that can act as an acyl donor. It can be synthesized into a series of pyrrolidine derivatives through tranesterification reactions with various alcohols.
  • Drug intermediate
  • Can act as an acyl donor
  • Can be synthesized into a series of pyrrolidine derivatives through tranesterification reactions with various alcohols
Catalog No. 50-004-52441 Supplier Medchemexpress LLC Supplier No. 25GHYW012802
Methyl 1H-pyrrole-2-carboxylate is a drug intermediate that can act as an acyl donor. It can be synthesized into a series of pyrrolidine derivatives through tranesterification reactions with various alcohols.
  • Drug intermediate
  • Can act as an acyl donor
  • Can be synthesized into a series of pyrrolidine derivatives through tranesterification reactions with various alcohols