Learn More
Medchemexpress LLC Methyl 2-(1H-indol-3-yl)acetate | 1912-33-0 | MFCD00022749 | 99.9% | 189.21 g·mol⁻1 | C11H11NO2 | 500 MG

Supplier: Medchemexpress LLC HYW015224500MG
Methyl 2-(1H-indol-3-yl)acetate is an endogenous indole-derived metabolite supplied as a research-grade chemical standard for analytical and metabolic studies. It is suitable as a reference standard, tracer, or reagent in biochemical assays and chromatographic analyses.
- Endogenous indole-derived metabolite.
- High purity (99.9%).
- Molecular formula C11H11NO2.
- Molecular weight 189.21 g·mol⁻1.
- Supplied in solid and solution formats.
- Storage: -20 °C (solid) and -80 °C (in solvent) for long-term stability.
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.