Learn More
Medchemexpress LLC Methyl 2-(1H-indol-3-yl)acetate | 1912-33-0 | MFCD00022749 | 99.9% | 189.21 g/mol | C11H11NO2 | 1 G
SDP
Catalog No. 5000303373
Encompass_Preferred
Methyl 2-(1H-indol-3-yl)acetate is an indole-derived methyl ester and endogenous metabolite supplied for research use. It is commonly used as a biochemical reagent in metabolic profiling, tracer studies, and analytical method development. The compound is available in both solid and solution formats and is provided with purity and storage specifications for laboratory use.
  • Used as a research reagent for metabolic profiling and biochemical studies.
  • Suitable for tracer and internal standard applications and analytical method development.
  • Available in solid and solution formats, including 1 g and small-volume solution packs.
  • High reported purity (99.9%).
  • Molecular formula C11H11NO2; molecular weight 189.21 g/mol.
  • Provided with recommended storage conditions for pure form and in solvent.
Catalog No. 50-003-03373 Supplier Medchemexpress LLC Supplier No. HYW0152241G
Methyl 2-(1H-indol-3-yl)acetate is an indole-derived methyl ester and endogenous metabolite supplied for research use. It is commonly used as a biochemical reagent in metabolic profiling, tracer studies, and analytical method development. The compound is available in both solid and solution formats and is provided with purity and storage specifications for laboratory use.
  • Used as a research reagent for metabolic profiling and biochemical studies.
  • Suitable for tracer and internal standard applications and analytical method development.
  • Available in solid and solution formats, including 1 g and small-volume solution packs.
  • High reported purity (99.9%).
  • Molecular formula C11H11NO2; molecular weight 189.21 g/mol.
  • Provided with recommended storage conditions for pure form and in solvent.