Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 2-(1H-indol-3-yl)acetate-d5 | 102415-39-4 | 99.1% | 194.24 | C11H6D5NO2 | 5 MG
SDP

Catalog No. 5000303667
Encompass_Preferred

Methyl 2-(1H-indol-3-yl)acetate-d5 is the deuterium-labeled analogue of the endogenous metabolite methyl 2-(1H-indol-3-yl)acetate. It is intended for research use as a stable-isotope reagent, commonly used as an internal standard or tracer in mass spectrometry and metabolic studies. The product is supplied with accompanying COA and SDS for analytical applications.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High chemical purity (99.08%).
  • High isotopic labeling for reliable quantitation.
  • Molecular formula C11H6D5NO2; molecular weight 194.24.
  • Supplied in small vial sizes suitable for analytical use.

Catalog No. 50-003-03667 Supplier Medchemexpress LLC Supplier No. HYW015224S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 2-(1H-indol-3-yl)acetate-d5 is the deuterium-labeled analogue of the endogenous metabolite methyl 2-(1H-indol-3-yl)acetate. It is intended for research use as a stable-isotope reagent, commonly used as an internal standard or tracer in mass spectrometry and metabolic studies. The product is supplied with accompanying COA and SDS for analytical applications.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High chemical purity (99.08%).
  • High isotopic labeling for reliable quantitation.
  • Molecular formula C11H6D5NO2; molecular weight 194.24.
  • Supplied in small vial sizes suitable for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.