Learn More
Cayman Chemical Methyl 2 1HIndazole3carbo 10mg
SDP
Catalog No. 503015943
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a precursor in the synthesis of synthetic cannabinoids including AMB, 5-fluoro AMB, MMB-4en-PINACA, MMB-FUBINACA, and MA-CHMINACA; a metabolite of 5-fluoro AMB and MA-CHMINACA; intended for research and forensic applications
Catalog No. 50-301-5943 Supplier Cayman Chemical Supplier No. 2558010 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a precursor in the synthesis of synthetic cannabinoids including AMB, 5-fluoro AMB, MMB-4en-PINACA, MMB-FUBINACA, and MA-CHMINACA; a metabolite of 5-fluoro AMB and MA-CHMINACA; intended for research and forensic applications