Learn More
Medchemexpress LLC Methyl 2-(2,4-dichlorophenoxy)acetate | 1928-38-7 | MFCD00018356 | ≥98.0% | 235.06 g/mol | C9H8Cl2O3 | 1G
SDP
Catalog No. 5000235503
Encompass_Preferred
Methyl 2-(2,4-dichlorophenoxy)acetate (2,4-D methyl ester; CAS 1928-38-7) is the methyl ester derivative of 2,4-dichlorophenoxyacetic acid supplied as an analytical standard and research reagent. It has chemical formula C9H8Cl2O3, molecular weight 235.06 g/mol, and is typically provided at ≥98.0% purity in gram-scale packaging for laboratory and analytical use. Supporting documentation commonly includes SDS, COA, NMR, and MS files.
  • Analytical standard for GC/MS and LC/MS methods.
  • High purity suitable for quantitative analysis.
  • Supplied in gram-scale quantities for laboratory experiments.
  • Includes SDS, certificate of analysis, and spectral data for verification.
  • IUPAC name provided for unambiguous chemical identification.
Catalog No. 50-002-35503 Supplier Medchemexpress LLC Supplier No. HY1245471G
Methyl 2-(2,4-dichlorophenoxy)acetate (2,4-D methyl ester; CAS 1928-38-7) is the methyl ester derivative of 2,4-dichlorophenoxyacetic acid supplied as an analytical standard and research reagent. It has chemical formula C9H8Cl2O3, molecular weight 235.06 g/mol, and is typically provided at ≥98.0% purity in gram-scale packaging for laboratory and analytical use. Supporting documentation commonly includes SDS, COA, NMR, and MS files.
  • Analytical standard for GC/MS and LC/MS methods.
  • High purity suitable for quantitative analysis.
  • Supplied in gram-scale quantities for laboratory experiments.
  • Includes SDS, certificate of analysis, and spectral data for verification.
  • IUPAC name provided for unambiguous chemical identification.