Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,2':6',2″-Terpyridine-4'-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M2464100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2464100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2464100MG Supplier TCI America Supplier No. M2464100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 247058-06-6
Molecular Formula C17H13N3O2
Molecular Weight (g/mol) 291.31
MDL Number MFCD11042546
InChI Key GEXSUZVSFVGSMZ-UHFFFAOYSA-N
Synonym 2,2′C:6′C,2′C′C-Terpyridine-4′C-carboxylic Acid Methyl Ester, 4′C-(Methoxycarbonyl)-2,2′C:6′C,2′C′C-terpyridine
PubChem CID 10589647
IUPAC Name methyl 2,6-dipyridin-2-ylpyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC(=C1)C2=CC=CC=N2)C3=CC=CC=N3

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 100 mg
Formula Weight 291.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,2′:6′,2′′-Terpyridine-4′-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.