Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl ™-(+)-2,2-Dimethyl-1,3-Dioxolane-4-Carboxylate

Catalog No. 3454825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-548-25G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-548-25G Supplier Sigma Aldrich Supplier No. 3454825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl ™-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate may be used as a starting material to synthesize (2R)-[1-2H2]-glycerol, a probe to study the stereochemistry of glycerol metabolism in Streptomyces cattleya. It may also be used to synthesize organic building blocks like ™-2,3-dihydroxy-3-methylbutyl toluene-p-sulfonate and (2S,8R)-2-amino-8-[™ 2,2-dimethyl-1,3-dioxolan-4-yl]-8-oxooctanoic acid.

Specifications

CAS 78761-37-2
Melting Point 150°C to 152°C
Molecular Formula C20H19NO7
Refractive Index n20/D 1.426
Linear Formula C20H19NO7
Quantity 50 g
Synonym alpha,beta-Isopropylidene-D-glyceric acid methyl ester; Methyl alpha,beta-isopropylidene-D-glycerate; Methyl 2,3-O-isopropylidene-D-glycerate
Molecular Weight (g/mol) 385.37 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.