Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 2-(2-oxoindolin-6-yl)acetate

Catalog No. SYV000011G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYV000011G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYV000011G Supplier Sigma Aldrich Supplier No. SYV000011G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methoxyphenylacetonitrile has been isolated from the sponge psammaplysilla purpurea.4-methoxyphenylacetonitrile was used as starting reagent in the synthesis of :
  • 2-(1-cyano-1-(2-methoxy)phenyl)methylidene-3-phenylthiazolidine-4,5-dione
  • 2-(1-cyano-1-(4-methoxy)-phenyl)methylidene-3-phenylthiazolidine-4,5-dione
  • α-cyanostilbenes

Specifications

CAS 14192-26-8
Molecular Formula C10H9NO3
Linear Formula C10H9NO3
MDL Number MFCD03095196
Quantity 100 g
Molecular Weight (g/mol) 191.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.