Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside 98.0+%, TCI America™
SDP

Catalog No. M16821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16821G 1 g
M16825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16821G Supplier TCI America Supplier No. M16821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13350-45-3
Molecular Formula C15H22O9S
Molecular Weight (g/mol) 378.392
InChI Key XWFUCHLBRWBKGN-QMIVOQANSA-N
PubChem CID 11164827
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-methylsulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)SC)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 91°C
Color White
Quantity 1 g
Formula Weight 378.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.