Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,3-dichloropropionate, 98%

Catalog No. p-7026156
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2199314 25 g
AAB2199322 100 g
AAB2199336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2199314 Supplier Thermo Scientific Chemicals Supplier No. B2199314
Only null left
Add to Cart

CAS: 3674-09-7 | C4H6Cl2O2 | 156.99 g/mol

Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.

Solubility
Soluble in ether, acetone, ethanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3674-09-7
Molecular Formula C4H6Cl2O2
Molecular Weight (g/mol) 156.99
MDL Number MFCD00000944
InChI Key OFHMODDLBXETIK-UHFFFAOYNA-N
IUPAC Name methyl 2,3-dichloropropanoate

Specifications

Density 1.326
Boiling Point 91°C to 92°C (50 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.453
Quantity 25 g
UN Number UN3265
Beilstein 1701891
Synonym methyl vinyl sulfone, ethene, methylsulfonyl, methyl vinyl sulphone, methylvinylsulfone, methanesulfonylethene, sulfone, methyl vinyl, vinyl methyl sulfone, methanesulfonyl-ethene, methylsulfonyl ethene, methyl vinyl sulfphone
Solubility Information Soluble in ether,acetone,ethanol.
SMILES COC(=O)C(Cl)CCl
PubChem CID 19354
Formula Weight 157
Percent Purity 98%
Chemical Name or Material Methyl 2,3-dichloropropionate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 222-940-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.