Learn More
Medchemexpress LLC Methyl 2,3-O-isopropylidene-5-O-tosyl-D-ribonucleoside | 4137-56-8 | 99.3% | 358.41 | 5 G
SDP
Catalog No. 5000404098
Encompass_Preferred
Methyl 2,3-O-isopropylidene-5-O-tosyl-D-ribonucleoside is a biochemical reagent that can be used for compound synthesis. It is for research use only.
  • Solid appearance
  • White to off-white color
  • Ships at room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-04098 Supplier Medchemexpress LLC Supplier No. HY244005G
Methyl 2,3-O-isopropylidene-5-O-tosyl-D-ribonucleoside is a biochemical reagent that can be used for compound synthesis. It is for research use only.
  • Solid appearance
  • White to off-white color
  • Ships at room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month