Learn More
Medchemexpress LLC Methyl 2,3-O-Isopropylidene-β-D-ribofuranoside | 4099-85-8 | 96.0% | 204.22 | 10 G
SDP
Catalog No. 5000436178
Encompass_Preferred
Methyl 2,3-O-Isopropylidene-β-D-ribofuranoside is a furanose derivative that serves as an important intermediate in the synthesis of various compounds.
  • Furanose derivative
  • Important intermediate for synthesis
  • Used in the synthesis of arsenic-containing ribosides
Catalog No. 50-004-36178 Supplier Medchemexpress LLC Supplier No. HY11394910G
Methyl 2,3-O-Isopropylidene-β-D-ribofuranoside is a furanose derivative that serves as an important intermediate in the synthesis of various compounds.
  • Furanose derivative
  • Important intermediate for synthesis
  • Used in the synthesis of arsenic-containing ribosides