Learn More
Medchemexpress LLC Methyl 2,3-O-Isopropylidene-β-D-ribofuranoside | 4099-85-8 | 96.0% | 204.22 | 25 G
SDP
Catalog No. 5000439911
Encompass_Preferred
Methyl 2,3-O-Isopropylidene-β-D-ribofuranoside is a furanose derivative. It serves as an important intermediate for the synthesis of compounds such as arsenic-containing ribosides.
  • Furanose derivative.
  • Important intermediate for the synthesis of arsenic-containing ribosides.
  • For research use only.
Catalog No. 50-004-39911 Supplier Medchemexpress LLC Supplier No. 25GHY113949
Methyl 2,3-O-Isopropylidene-β-D-ribofuranoside is a furanose derivative. It serves as an important intermediate for the synthesis of compounds such as arsenic-containing ribosides.
  • Furanose derivative.
  • Important intermediate for the synthesis of arsenic-containing ribosides.
  • For research use only.