Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 2,3-O-Isopropylidene-β-L-ribofuranoside | 20672-63-3 | 97.1% | 204.22 | 1 ML
SDP

Catalog No. 5000436758
Encompass_Preferred

Methyl 2,3-O-Isopropylidene-β-L-ribofuranoside is an enantiomer of Methyl 2,3-O-Isopropylidene-β-D-ribofuranoside and a derivative of L-ribose. It is intended for research use only.

  • Purity of 97.10%
  • Molecular weight of 204.22
  • Chemical formula C9H16O5
  • CAS number 20672-63-3
  • Appearance as a colorless to light yellow liquid
  • In vitro solubility of 100 mg/mL in DMSO
  • Recommended storage at 4°C, protected from light, and under nitrogen

Catalog No. 50-004-36758 Supplier Medchemexpress LLC Supplier No. HY113949A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 2,3-O-Isopropylidene-β-L-ribofuranoside is an enantiomer of Methyl 2,3-O-Isopropylidene-β-D-ribofuranoside and a derivative of L-ribose. It is intended for research use only.

  • Purity of 97.10%
  • Molecular weight of 204.22
  • Chemical formula C9H16O5
  • CAS number 20672-63-3
  • Appearance as a colorless to light yellow liquid
  • In vitro solubility of 100 mg/mL in DMSO
  • Recommended storage at 4°C, protected from light, and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.