Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 2-(4-bromophenyl)-6-phenylpyridine-4-carboxylate

Catalog No. JRD10901G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD10901G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD10901G Supplier Sigma Aldrich Supplier No. JRD10901G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(tert-Butoxycarbonyl)-2-bromoaniline, also known as tert-butyl-N-(2-bromophenyl)carbamate, is an N-Boc-protected o-bromoaniline.1 It reacts with ethyl perfluorooctanoate in the presence of tert-butyllithium to form the corresponding 1-hydroxy-1H-perfluorooctyl compound.N-(tert-Butoxycarbonyl)-2-bromoaniline may be used in the synthesis of N-boc-protected o-aryl anilines, which undergoes coupling reaction with bromoacetylenes to form the corresponding arene?ynamides.

Specifications

Molecular Formula C19H14BrNO2
Linear Formula C19H14BrNO2
MDL Number MFCD12196784
Quantity 10 g
Molecular Weight (g/mol) 368.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.