Learn More
Medchemexpress LLC Methyl 2-[(4-chlorobenzoyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate | 544450-68-2 | 99.9% | C16H16ClNO3S | 10MG
SDP
Catalog No. 5000260202
Encompass_Preferred
COR659 (methyl 2-[(4-chlorobenzoyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate; CAS 544450-68-2) is a small-molecule positive allosteric modulator of the GABAB receptor used in preclinical neuroscience and addiction research. It has been reported to reduce alcohol- and chocolate-seeking behavior in rodent models and is supplied as a high-purity research chemical for in vitro and in vivo pharmacology studies.
  • Potent GABAB positive allosteric modulator with demonstrated in vivo activity.
  • High purity suitable for research applications.
  • Well characterized chemical identifiers and molecular formula.
  • Off-white solid form compatible with standard solvents such as DMSO.
  • Storage and handling guidance provided for refrigerated and cryogenic conditions.
Catalog No. 50-002-60202 Supplier Medchemexpress LLC Supplier No. HY13720410MG
COR659 (methyl 2-[(4-chlorobenzoyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate; CAS 544450-68-2) is a small-molecule positive allosteric modulator of the GABAB receptor used in preclinical neuroscience and addiction research. It has been reported to reduce alcohol- and chocolate-seeking behavior in rodent models and is supplied as a high-purity research chemical for in vitro and in vivo pharmacology studies.
  • Potent GABAB positive allosteric modulator with demonstrated in vivo activity.
  • High purity suitable for research applications.
  • Well characterized chemical identifiers and molecular formula.
  • Off-white solid form compatible with standard solvents such as DMSO.
  • Storage and handling guidance provided for refrigerated and cryogenic conditions.