Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-Dichlorobenzoate 98.0+%, TCI America™
SDP

Catalog No. D28555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28555G Supplier TCI America Supplier No. D28555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35112-28-8
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00085117
InChI Key VCRWILYAWSRHBN-UHFFFAOYSA-N
Synonym methyl2,4-dichlorobenzoate, 2,4-dichlorobenzoic acid methyl ester, benzoic acid, 2,4-dichloro-, methyl ester, methyl-2,4-dichlorobenzoate, pubchem3713, methyl ester of 2,4-dichlorobenzoic acid, acmc-209id8, ksc497m5b, methyl 2,4-dichloro benzoate
PubChem CID 37055
IUPAC Name methyl 2,4-dichlorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)Cl)Cl

Specifications

Color Yellow
Boiling Point 239°C
Quantity 5 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,4-Dichlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.