Learn More
Medchemexpress LLC Methyl 2,4-dichlorophenoxyacetate | 1928-38-7 | MFCD00018356 | ≥98.0% | 235.06 g/mol | C9H8Cl2O3 | 10G
SDP
Catalog No. 5000237823
Encompass_Preferred
2,4-D methyl ester (methyl 2,4-dichlorophenoxyacetate) is the methyl ester derivative of 2,4-dichlorophenoxyacetic acid. Supplied as a research-grade analytical reagent for laboratory use, it is commonly used as an analytical standard and reference compound in environmental and biochemical analyses. Solid form: C9H8Cl2O3; CAS 1928-38-7; MW 235.06 g/mol.
  • Purity ≥98.0%.
  • Available in 1 g, 5 g, and 10 g packaging.
  • Suitable as an analytical standard for pesticide and environmental testing.
  • Solid form for stable storage and handling.
  • Certificate of analysis available for each batch.
Catalog No. 50-002-37823 Supplier Medchemexpress LLC Supplier No. HY12454710G
2,4-D methyl ester (methyl 2,4-dichlorophenoxyacetate) is the methyl ester derivative of 2,4-dichlorophenoxyacetic acid. Supplied as a research-grade analytical reagent for laboratory use, it is commonly used as an analytical standard and reference compound in environmental and biochemical analyses. Solid form: C9H8Cl2O3; CAS 1928-38-7; MW 235.06 g/mol.
  • Purity ≥98.0%.
  • Available in 1 g, 5 g, and 10 g packaging.
  • Suitable as an analytical standard for pesticide and environmental testing.
  • Solid form for stable storage and handling.
  • Certificate of analysis available for each batch.