Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-Dichlorophenylacetate 99.0+%, TCI America™
SDP

Catalog No. M25285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25285G 5 g
M252825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25285G Supplier TCI America Supplier No. M25285G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55954-23-9
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.061
MDL Number MFCD00018381
InChI Key SRCVNBFTEARRJY-UHFFFAOYSA-N
Synonym methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, 2,4-dichlorophenylacetic acid methyl ester, benzeneacetic acid, 2,4-dichloro-, methyl ester, methyl2,4-dichlorophenylacetate, dcaa methyl ester, acmc-209lq7, methyl 2,4-dichlorobenzeneacetate, 2,4-dichlorophenylaceticacidmethylester, 2,4-dichlorobenzeneacetic acid methyl ester
PubChem CID 2734106
IUPAC Name methyl 2-(2,4-dichlorophenyl)acetate
SMILES COC(=O)CC1=C(C=C(C=C1)Cl)Cl

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 g
Formula Weight 219.06
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,4-Dichlorophenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.