Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-Difluorobenzoate 98.0+%, TCI America™
SDP

Catalog No. M18795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18795G 5 g
M187925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18795G Supplier TCI America Supplier No. M18795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106614-28-2
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD03093799
InChI Key YQUHULOBTDYMAG-UHFFFAOYSA-N
Synonym 2,4-difluoro-benzoic acid methyl ester, 2,4-difluorobenzoic acid methyl ester, methyl-2,4-difluorobenzoate, methyl 2,4-difluoro benzoate, benzoic acid, 2,4-difluoro-, methyl ester, pubchem15678, acmc-1c4sx, methyl 2,4-diflourobenzoate, ksc494g3p, rarechem al bf 0213
PubChem CID 2782897
IUPAC Name methyl 2,4-difluorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)F)F

Specifications

Color Yellow
Boiling Point 194°C
Quantity 5 g
Formula Weight 172.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,4-Difluorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.