Learn More
Medchemexpress LLC Methyl 2,4-dihydroxybenzoate | 2150-47-2 | 99.6% | 168.15 | 250 MG
SDP
Catalog No. 5000366406
Encompass_Preferred
Methyl 2,4-dihydroxybenzoate is a key intermediate that can be utilized in the formation of other active compounds. It is intended for research use only and is not sold to patients. The product appears as a solid with a color ranging from white to pink. It should be stored at room temperature in the continental US, and at 4°C under nitrogen. For solutions, storage at -80°C for 6 months or -20°C for 1 month (under nitrogen) is recommended.
  • Key intermediate for forming other active compounds.
  • Available in various quantities (e.g., 100 mg, 250 mg, 500 mg, 1 g).
  • Can be purchased as a solid or in solution (10 mM * 1 mL in DMSO).
  • High purity: 99.57%.
  • Neuroprotective activity observed in human SH-SY5Y cells.
Catalog No. 50-003-66406 Supplier Medchemexpress LLC Supplier No. 250MGHYW010076
Methyl 2,4-dihydroxybenzoate is a key intermediate that can be utilized in the formation of other active compounds. It is intended for research use only and is not sold to patients. The product appears as a solid with a color ranging from white to pink. It should be stored at room temperature in the continental US, and at 4°C under nitrogen. For solutions, storage at -80°C for 6 months or -20°C for 1 month (under nitrogen) is recommended.
  • Key intermediate for forming other active compounds.
  • Available in various quantities (e.g., 100 mg, 250 mg, 500 mg, 1 g).
  • Can be purchased as a solid or in solution (10 mM * 1 mL in DMSO).
  • High purity: 99.57%.
  • Neuroprotective activity observed in human SH-SY5Y cells.