Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 2-(4-fluorophenyl)-6-(4-methoxyphenyl)pyridine-4-carboxylate

Catalog No. JRD11051G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD11051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD11051G Supplier Sigma Aldrich Supplier No. JRD11051G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-(1h-pyrrol-1-yl)ethanamine may be used in the preparation of dihydropyrrolo[2',1':3,4]pyrazino[2,1-a]isoindolones by reacting with 2-formylbenzoic acids or 2-acetylbenzoic acid via n-acyliminium cation aromatic cyclizations
  • This product was previously listed as cbr00085.convenient syntheses of dihydropyrrolo [2',1':3,4] pyrazino-and dihydropyrrolo [2',1':3,4][1,4] diazepino-[2,1-a] isoindolones

Specifications

Molecular Formula C20H16FNO3
Linear Formula C20H16FNO3
MDL Number MFCD12196799
Quantity 1 g
Molecular Weight (g/mol) 337.34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.