Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-[4-(Hydroxymethyl)-2-nitrophenoxy]acetate 98.0+%, TCI America™
SDP

Catalog No. H10491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10491G 1 g
H1049100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10491G Supplier TCI America Supplier No. H10491G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 308815-81-8
Molecular Formula C10H11NO6
Molecular Weight (g/mol) 241.20
MDL Number MFCD04117920
InChI Key HNFVHTJFCGBTSS-UHFFFAOYSA-N
Synonym 2-[4-(Hydroxymethyl)-2-nitrophenoxy]acetic Acid Methyl Ester
PubChem CID 43137771
IUPAC Name methyl 2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate
SMILES COC(=O)COC1=C(C=C(CO)C=C1)[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 241.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-[4-(Hydroxymethyl)-2-nitrophenoxy]acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.