Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,5-Bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) 95.0+%, TCI America™
SDP

Catalog No. M20305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20305G Supplier TCI America Supplier No. M20305G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26131-32-8
Molecular Formula C10H3F17O4
Molecular Weight (g/mol) 510.103
MDL Number MFCD00054723
InChI Key UMADKHFVSXRTGY-UHFFFAOYSA-N
Synonym methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-perfluoropropoxy propoxy propanoate, perfluoro-2,5-dimethyl-3,6-dioxanonanoic acid methyl ester, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propanoate, methyl 2,3,3,3-tetrafluoro-2-1,1,2,3,3,3-hexafluoro-2-heptafluoropropoxy propoxy propanoate, cf3cf2cf2ocf cf3 cf2ocf cf3 cooch3, methyl perfluoro-2,5-dimethyl-3,6-dioxa-nonanoate, methyl 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate, 2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoic acid methyl ester, 2-2-heptafluoropropoxy hexafluoropropoxy-2,3,3,3-tetrafluoropropionic acid methyl ester, methyl2,5-bis trifluoromethyl-3,6-dioxaundecafluorononanoate mixtureofisomers
PubChem CID 141268
IUPAC Name methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate
SMILES COC(=O)C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 g
Formula Weight 510.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,5-Bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.