Learn More
Medchemexpress LLC Methyl 2,5-dihydroxycinnamate | 63177-57-1 | MFCD00132932 | 99.8% | 194.18 g/mol | C10H10O4 | 25 MG
SDP
Catalog No. 5000211897
Encompass_Preferred
Methyl 2,5-dihydroxycinnamate is a methyl cinnamate derivative that acts as an erbstatin analog and a potent, stable inhibitor of epidermal growth factor receptor (EGFR) kinase activity. It is supplied for research use in solid form and as DMSO solutions, and is intended for biochemical and cellular assays to probe EGFR-related signaling. Handle according to laboratory safety practices.
  • Potent EGFR kinase inhibitor for signaling studies.
  • High purity (99.8%) suitable for sensitive assays.
  • Molecular weight 194.18 g/mol and formula C10H10O4 for accurate calculations.
  • Available in small solid quantities and as 10 mM DMSO solution for convenient dosing.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-11897 Supplier Medchemexpress LLC Supplier No. HY10100625MG
Add to Cart
Add to Cart
Methyl 2,5-dihydroxycinnamate is a methyl cinnamate derivative that acts as an erbstatin analog and a potent, stable inhibitor of epidermal growth factor receptor (EGFR) kinase activity. It is supplied for research use in solid form and as DMSO solutions, and is intended for biochemical and cellular assays to probe EGFR-related signaling. Handle according to laboratory safety practices.
  • Potent EGFR kinase inhibitor for signaling studies.
  • High purity (99.8%) suitable for sensitive assays.
  • Molecular weight 194.18 g/mol and formula C10H10O4 for accurate calculations.
  • Available in small solid quantities and as 10 mM DMSO solution for convenient dosing.
  • For research use only; not for human or clinical use.