Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,6-dichloropyridine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAA1746806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1746806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1746806 Supplier Thermo Scientific Chemicals Supplier No. A1746806

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates.

Solubility
Sparingly Soluble (0.13 g/L) (25°C)

Notes
Keep away from oxidizing agents, reducing agents, bases, amines. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42521-09-5
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00044409
InChI Key XSKGHSUHOYEBTK-UHFFFAOYSA-N
PubChem CID 93237
IUPAC Name methyl 2,6-dichloropyridine-4-carboxylate
SMILES COC(=O)C1=CC(Cl)=NC(Cl)=C1

Specifications

Melting Point 80°C to 84°C
Quantity 5 g
Synonym methyl 2,6-dichloroisonicotinate, methyl2,6-dichloroisonicotinate, 2,6-chloro-4-pyridinecarboxylic acid methyl ester, 2,6-dichloroisonicotinic acid methyl ester, methyl-2,6-dichloroisonicotinate, 4-pyridinecarboxylic acid, 2,6-dichloro-, methyl ester, methyl-2,6-dichlorisonicotinat, acmc-209yt4, xskghsuhoyebtk-uhfffaoysa, methyl 2,6-dichloro-isonicotinate
Solubility Information Sparingly Soluble (0.13g/L) (25°C)
Formula Weight 206.03
Percent Purity 97%
Chemical Name or Material Methyl 2,6-dichloropyridine-4-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 255-868-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.