Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 2-((6S)-4-(4-chlorophenyl)-2-((5-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino) | 2243076-67-5 | 98.9% | 50 MG
SDP

Catalog No. 5000231984
Encompass_Preferred

ZXH-3-26 is a proteolysis targeting chimera (PROTAC) that links a Cereblon ligand to a BRD4 ligand to induce selective BRD4 degradation, with a reported DC50/5h of approximately 5 nM. It is supplied as a 50 mg vial at high purity for use in chemical biology and targeted protein degradation research.

  • Selective BRD4 degradation with DC50/5h ≈ 5 nM.
  • Minimal degradation of BRD2 and BRD3 at tested concentrations.
  • Molecular formula C38H37ClN8O7S and molecular weight 785.27.
  • Purity 98.9% and supplied as a 50 mg quantity.
  • Includes SMILES for cheminformatics and structure confirmation.

Catalog No. 50-002-31984 Supplier Medchemexpress LLC Supplier No. HY12282650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZXH-3-26 is a proteolysis targeting chimera (PROTAC) that links a Cereblon ligand to a BRD4 ligand to induce selective BRD4 degradation, with a reported DC50/5h of approximately 5 nM. It is supplied as a 50 mg vial at high purity for use in chemical biology and targeted protein degradation research.

  • Selective BRD4 degradation with DC50/5h ≈ 5 nM.
  • Minimal degradation of BRD2 and BRD3 at tested concentrations.
  • Molecular formula C38H37ClN8O7S and molecular weight 785.27.
  • Purity 98.9% and supplied as a 50 mg quantity.
  • Includes SMILES for cheminformatics and structure confirmation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.