Learn More
Medchemexpress LLC Methyl 2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetate | 15991-13-6 | 97.4% | 234.20 g·mol⁻¹ | C12H10O5 | 1 ML
SDP
Catalog No. 5000301101
Encompass_Preferred
Methyl 2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetate is a coumarin-derived small-molecule fluorescent compound supplied as a solid or as a ready-to-use 10 mM solution in DMSO. It has a molecular weight of 234.20 g·mol⁻¹ and CAS number 15991-13-6. It is commonly used in fluorescence-based assays and synthetic chemistry where a coumarin fluorophore is required.
  • Coumarin-derived fluorescent core for strong fluorescence.
  • Available as solid or 10 mM solution in DMSO for convenient use.
  • High purity (~97%) suitable for analytical and preparative work.
  • Low molecular weight for efficient incorporation into small-molecule probes.
  • White to light yellow solid appearance for easy visual inspection.
Catalog No. 50-003-01101 Supplier Medchemexpress LLC Supplier No. HYW03031910MM/1ML
Methyl 2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetate is a coumarin-derived small-molecule fluorescent compound supplied as a solid or as a ready-to-use 10 mM solution in DMSO. It has a molecular weight of 234.20 g·mol⁻¹ and CAS number 15991-13-6. It is commonly used in fluorescence-based assays and synthetic chemistry where a coumarin fluorophore is required.
  • Coumarin-derived fluorescent core for strong fluorescence.
  • Available as solid or 10 mM solution in DMSO for convenient use.
  • High purity (~97%) suitable for analytical and preparative work.
  • Low molecular weight for efficient incorporation into small-molecule probes.
  • White to light yellow solid appearance for easy visual inspection.