Learn More
Medchemexpress LLC Methyl 2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetate | 15991-13-6 | MFCD01463811 | 97.4% | 234.20 g/mol | C12H10O5 | 25 MG
SDP
Catalog No. 5000303848
Encompass_Preferred
Methyl 2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetate is a coumarin-class small molecule used as a fluorescent probe and synthetic intermediate in chemical and biological research. Supplied as a solid with high listed purity, it is intended for analytical assays, fluorescence-based applications, and lab-scale synthesis.
  • Solid, white to light yellow appearance.
  • High purity suitable for analytical and synthetic use.
  • Coumarin-derived structure with fluorescent properties.
  • Available in multiple milligram packaging sizes for lab-scale work.
  • Provided with standard chemical identifiers for reproducibility.
Catalog No. 50-003-03848 Supplier Medchemexpress LLC Supplier No. 25MGHYW030319
Methyl 2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetate is a coumarin-class small molecule used as a fluorescent probe and synthetic intermediate in chemical and biological research. Supplied as a solid with high listed purity, it is intended for analytical assays, fluorescence-based applications, and lab-scale synthesis.
  • Solid, white to light yellow appearance.
  • High purity suitable for analytical and synthetic use.
  • Coumarin-derived structure with fluorescent properties.
  • Available in multiple milligram packaging sizes for lab-scale work.
  • Provided with standard chemical identifiers for reproducibility.