Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Acetamidopropionate 98.0+%, TCI America™
SDP

Catalog No. A143425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A143425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A143425G Supplier TCI America Supplier No. A143425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26629-33-4
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.16
MDL Number MFCD01631289
InChI Key FQGVVDYNRHNTCK-UHFFFAOYNA-N
Synonym 2-Acetamidopropionic Acid Methyl Ester, N-Acetyl-DL-alanine Methyl Ester
PubChem CID 520278
IUPAC Name methyl 2-acetamidopropanoate
SMILES COC(=O)C(C)NC(C)=O

Specifications

Melting Point 48°C
Color White
Quantity 25 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Acetamidopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.