Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Amino-4-(trifluoromethyl)benzoate 98.0+%, TCI America™
SDP

Catalog No. M29055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29055G 5 g
M29051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29055G Supplier TCI America Supplier No. M29055G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61500-87-6
Molecular Formula C9H8F3NO2
Molecular Weight (g/mol) 219.163
MDL Number MFCD08543939
InChI Key DZICUHOFOOPVFM-UHFFFAOYSA-N
Synonym 2-Amino-4-(trifluoromethyl)benzoic Acid Methyl Ester
PubChem CID 12601886
IUPAC Name methyl 2-amino-4-(trifluoromethyl)benzoate
SMILES COC(=O)C1=C(C=C(C=C1)C(F)(F)F)N

Specifications

Melting Point 63°C
Quantity 5 g
Formula Weight 219.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Amino-4-(trifluoromethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.