Learn More
Advanced Chemblocks Inc Methyl 2-amino-6-chloronicotinate; 1G; 97%
Catalog No. 501099993
Encompass
Chemical Name: Methyl 2-amino-6-chloronicotinate CAS: 849805-25-0 MW: 186.6 MDL Number: MFCD17015967 Category: pyridine Synonym: Methyl 2-amino-6-chloronicotinate
Catalog No. 50-109-9993 Supplier Advanced Chemblocks Inc Supplier No. C23581G
Chemical Name: Methyl 2-amino-6-chloronicotinate CAS: 849805-25-0 MW: 186.6 MDL Number: MFCD17015967 Category: pyridine Synonym: Methyl 2-amino-6-chloronicotinate