Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 2-aminoisobutyrate hydrochloride | 15028-41-8 | 153.61 | 250 G
SDP

Catalog No. 5000450022
Encompass_Preferred

Methyl 2-aminoisobutyrate hydrochloride is a biochemical reagent suitable for various life science related research. It can be utilized as a biological material or an organic compound. This product is intended for research use only.

  • Biochemical reagent for life science research
  • Can be used as a biological material
  • Can be used as an organic compound
  • Appearance: Solid, white to off-white
  • Storage: -20°C, sealed, away from moisture
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-50022 Supplier Medchemexpress LLC Supplier No. 250GHYW002520
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 2-aminoisobutyrate hydrochloride is a biochemical reagent suitable for various life science related research. It can be utilized as a biological material or an organic compound. This product is intended for research use only.

  • Biochemical reagent for life science research
  • Can be used as a biological material
  • Can be used as an organic compound
  • Appearance: Solid, white to off-white
  • Storage: -20°C, sealed, away from moisture
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.