Learn More
Medchemexpress LLC Methyl 2-aminonicotinate | 14667-47-1 | MFCD00128872 | 99.9% | C7H8N2O2 | 25 G

Supplier: Medchemexpress LLC 25GHY76735
Methyl 2-aminonicotinate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Used as a biological material or organic compound
- Suitable for life science related research
- Appears as a solid
- Color is white to yellow
- Store at 4°C, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.